Arsenic(III) oxide, 99.99% (metals basis)
Arsenic(III) oxide, 99.99% (metals basis)
Arsenic(III) oxide, 99.99% (metals basis)
Arsenic(III) oxide, 99.99% (metals basis)
Thermo Scientific Chemicals
Arsenic(III) oxide, 99.99% (metals basis)
CAS: 1327-53-3 | As2O3 | 197.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number ALF040370.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Boiling Point465°C (sublimation)
Chemical Name or MaterialArsenic(III) oxide
Melting Point312.3°C
CAS1327-53-3
ColorWhite
View more
used in the manufacturing of such products as glass and enamels. Arsenic trioxide is utilized in various studies on cellular processes, notably in investigations of leukemia and principal material for the production of many arsenic compounds.