Cerium molybdenum oxide, 99%
Cerium molybdenum oxide, 99%
Cerium molybdenum oxide, 99%
Cerium molybdenum oxide, 99%
Thermo Scientific Chemicals
Cerium molybdenum oxide, 99%
CAS: 13454-70-1 | Ce2Mo3O12 | 760.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF040210.065 g
Catalog number ALF040210.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium molybdenum oxide
Melting Point973°C
CAS13454-70-1
ColorBrown
Recommended StorageAmbient temperatures
View more
Cerium molybdenum oxide is used in the aerospace and coating industries.