Altus™ Table System Component, Service Assembly
Altus™ Table System Component, Service Assembly
Actual product may vary
Thermo Scientific Chemicals
Altus™ Table System Component, Service Assembly
Mott service assemblies have epoxy coated fixtures use with Altus table system
Have Questions?
Catalog NumberMaterialColorLocationOptions Available
ALF040001.22White hoseBlackZone 7Service: HE
Catalog number ALF040001.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
White hose
Color:
Black
Location:
Zone 7
Options Available:
Service: HE
  • Assembly complete with panel mounted single service fixture (Needle Valve supplied), keyed quick connect, and pug hose. Note: Cold water is supplied with ball valve
  • Corrugated gas appliance connector with stainless steel keyed quick connect is supplied for natural gas
  • Supplied with 1.2m (4 ft.) of service lines above table height
  • Epoxy coated fixtures
  • Fixtures may not be exactly as shown
  • Only plug and body quick connects of the same color will couple and allow flow
  • Parts are shipped loose
Order Destinations
  • Zone 1 (Use CC suffix): Montana (Eastern), Minnesota, North Dakota, South Dakota, Wyoming
  • Zone 2 (Use WJ suffix): Iowa, Kansas, Missouri, Nebraska, Wisconsin
  • Zone 3 (Use SL suffix): Alabama, Florida, Georgia, Illinois, Kentucky, Tennessee
  • Zone 4 (Use NE suffix): Connecticut, Delaware, District of Columbia, Maine, Maryland, Massachusetts, New Hampshire, North Carolina, Rhode Island, South Carolina, Vermont, Virginia, West Virginia
  • Zone 5 (Previous DT suffix): Moved to Zone 9
  • Zone 6 (Previous CF suffix): Moved to Zone 9
  • Zone 7 (Use MGC suffix): Arizona, Arkansas, Colorado, Louisiana, Mississippi, New Mexico, Oklahoma, Texas
  • Zone 8 (Use S suffix): New Jersey, New York, Pennsylvania (Eastern)
  • Zone 9 (Use SV suffix): Michigan, New York State (Upper), Ohio, Pennsylvania (Western)
  • Zone 10 (Use SB suffix): Alaska, California, Hawaii, Idaho, Montana (Western), Nevada, Oregon, Utah, Washington
Specifications
For Use With (Equipment)Altus™ Table System
LocationZone 7
Options AvailableService: HE
Product TypeService Assembly
ColorBlack
MaterialWhite hose
Unit SizeEA