Aluminum oxide, polishing compound
Aluminum oxide, polishing compound
Aluminum oxide, polishing compound
Aluminum oxide, polishing compound
Thermo Scientific Chemicals
Aluminum oxide, polishing compound
CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Catalog NumberQuantity
ALF039807.LA57 g
Catalog number ALF039807.LA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
57 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum oxide
OdorOdorless
Particle Size0.3 Micron
Physical FormParticles
Quantity57 g
View more
Polishing compound for IR crystal windows