Acetic acid, glacial, 99.7+% (metals basis)
Acetic acid, glacial, 99.7+% (metals basis)
Acetic acid, glacial, 99.7+% (metals basis)
Acetic acid, glacial, 99.7+% (metals basis)
Thermo Scientific Chemicals
Acetic acid, glacial, 99.7+% (metals basis)
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Catalog NumberQuantity
ALF039745.AE100 mL
Catalog number ALF039745.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic Acid
Name NoteGlacial
pH2.5
CAS64-19-7
Melting Point16.6°C
View more
Good polar protic solvent, in its ester form it act as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.