Carbon black, acetylene, 50% compressed, 99.9+%
Carbon black, acetylene, 50% compressed, 99.9+%
Actual product may vary
Carbon black, acetylene, 50% compressed, 99.9+%
Carbon black, acetylene, 50% compressed, 99.9+%
Thermo Scientific Chemicals
Carbon black, acetylene, 50% compressed, 99.9+%
Have Questions?
Catalog NumberQuantity
ALF039724.1850 g
Catalog number ALF039724.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Specifications
Quantity50 g
Percent Purity≥99.9%
Unit SizeEA