Sulfuric acid, 50% v/v aq. soln.
Sulfuric acid, 50% v/v aq. soln.
Sulfuric acid, 50% v/v aq. soln.
Sulfuric acid, 50% v/v aq. soln.
Thermo Scientific Chemicals
Sulfuric acid, 50% v/v aq. soln.
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Catalog NumberQuantity
ALF039668.K21 L
Catalog number ALF039668.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSulfuric acid, 50% v/v aq. soln.
CAS7664-93-9
Health Hazard 1H290-H314-H335
Health Hazard 3P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c