Tin(II) trifluoromethanesulfonate, 97%
Tin(II) trifluoromethanesulfonate, 97%
Tin(II) trifluoromethanesulfonate, 97%
Tin(II) trifluoromethanesulfonate, 97%
Thermo Scientific Chemicals
Tin(II) trifluoromethanesulfonate, 97%
CAS: 62086-04-8 | CH4F3O3SSn+ | 271.805 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF039603.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTin(II) trifluoromethanesulfonate
CAS62086-04-8
Health Hazard 1H314-H500
Health Hazard 2GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Tin(II) trifluoromethanesulfonate is a mild Lewis acid used for stereoselective aldol and Michael reactions; [2,3]-Wittig and Ireland-Claisen rearrangements, Horner-Wadsworth-Emmons reactions, asymmetric [3 + 2] cycloaddition, ring opening reactions and Friedel-Crafts reactions.