Boron, plasma standard solution, Specpure™ B 1000μg/mL
Boron, plasma standard solution, Specpure™ B 1000μg/mL
Boron, plasma standard solution, Specpure™ B 1000μg/mL
Boron, plasma standard solution, Specpure™ B 1000μg/mL
Thermo Scientific Chemicals
Boron, plasma standard solution, Specpure™ B 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF039147.AE100 mL
Catalog number ALF039147.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBoron
Name Noteplasma standard solution, B 1000μg/mL
Health Hazard 1H360FD
Health Hazard 3P201-P202-P281-P308+P313-P501c