Dichloromethane, ACS, 99.5+%, stab. with amylene
Dichloromethane, ACS, 99.5+%, stab. with amylene
Dichloromethane, ACS, 99.5+%, stab. with amylene
Dichloromethane, ACS, 99.5+%, stab. with amylene
Thermo Scientific Chemicals
Dichloromethane, ACS, 99.5+%, stab. with amylene
Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.002%, Titratable acid 0.0003meq/g, Free halogens P.T., H2O 0.02% | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Catalog NumberQuantity
ALF039116.K42.5 L
Catalog number ALF039116.K4
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Chemical Name or MaterialDichloromethane
CAS75-09-2
ColorColorless
Health Hazard 1H315-H319-H336-H350-H373
Health Hazard 3P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
View more
Dichloromethane is a chlorinated hydrocarbon that has been employed as a pharamacuetical applications. Mainly, it's utilization as a solvent in manufacturing and food engineering. It is too useful in the drinking bird heat engine. It was the solvent of choice for caffeine extraction. It is likewise employed as a solvent and paint remover. It is useful for the fraction of flash pyrolysis oil and for the extraction of benzo-15-crown-5.