Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL
Thermo Scientific Chemicals
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF038728.AE100 mL
Catalog number ALF038728.AE
Price (MYR)
413.00
EA
Add to cart
Quantity:
100 mL
Request bulk or custom format
Price (MYR)
413.00
EA
Add to cart
Specifications
Chemical Name or MaterialChromium plasma standard solution
Name NoteCr 1000μg/mL
Health Hazard 1H290-H314-H317-H332-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c