Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml
CAS: 7784-27-2 | AlH18N3O18 | 375.129 g/mol
Have Questions?
Change view
Quantity:
500 mL
100 mL
Catalog number ALF038727.AP
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
Name Noteplasma standard solution, Al 1000μg/mL
CAS7784-27-2
Health Hazard 1H290-H314-H335
CAS7697-37-2