Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
Thermo Scientific Chemicals
Buffer solution pH 4.00 (+/-0.022 @ 25oC) Colored Red Specpure NIST Traceable
CAS: 1310-73-2
Have Questions?
Catalog NumberQuantity
ALF038713.AP500 mL
Catalog number ALF038713.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS1310-73-2, 7778-77-0, 7732-18-5
Specifications
Chemical Name or MaterialBuffer solution
CAS1310-73-2
ColorRed
CAS7778-77-0
Recommended StorageAmbient temperatures
View more
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.