Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Methyl Red, 0.1% w/v solution in ethanol
Thermo Scientific Chemicals
Methyl Red, 0.1% w/v solution in ethanol
CAS: 74886-84-3
Have Questions?
Catalog NumberQuantity
ALF038698.AP500 mL
Catalog number ALF038698.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl Red
CAS74886-84-3
Health Hazard 1H225-H302-H319-H331-H370
Health Hazard 2GHS H Statement
H224
Extremely flammable liquid and vapor.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P311-P330-P370+P378q-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use