Platinum foil, 0.25mm (0.01in) thick, 99.95% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, 99.95% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, 99.95% (metals basis)
Platinum foil, 0.25mm (0.01in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals
Platinum foil, 0.25mm (0.01in) thick, 99.95% (metals basis)
CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF038378.FF25 x 25 mm
Catalog number ALF038378.FF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialPlatinum foil
Name Note0.25mm (0.01 in.) thick
CAS7440-06-4
Recommended StorageAmbient temperatures
EINECS Number231-116-1