Rosin
Rosin
Rosin
Thermo Scientific Chemicals
Rosin
CAS: 8050-09-7 | C20H30O2 | 302.46 g/mol
Have Questions?
Catalog NumberQuantity
ALF036734.30250 g
Catalog number ALF036734.30
Price (MYR)
308.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
308.00
EA
Add to cart
Specifications
Chemical Name or MaterialRosin
CAS8050-09-7
Health Hazard 1H317
Health Hazard 2GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3P261-P272-P280g-P302+P352-P333+P313-P363-P501c
View more
Rosin is an ingredient in printing inks, photocopying and laser printing paper, varnishes, adhesives (glues), soap, paper sizing, soda, soldering fluxes and sealing wax. It can be used as a glazing agent in medicines and chewing gum. In industry, rosin is a flux used in soldering. A mixture of pitch and rosin is used to make a surface against which glass is polished when making optical components such as lenses.