Bismuth 2-ethylhexanoate
Bismuth 2-ethylhexanoate
Bismuth 2-ethylhexanoate
Bismuth 2-ethylhexanoate
Thermo Scientific Chemicals
Bismuth 2-ethylhexanoate
CAS: 67874-71-9 | C24H48BiO6 | 641.622 g/mol
Have Questions?
Catalog NumberQuantity
ALF036710.1425 g
Catalog number ALF036710.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBismuth 2-ethylhexanoate
CAS67874-71-9
Health Hazard 1H361d
Health Hazard 2GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Health Hazard 3P201-P202-P281-P308+P313-P501c
View more
Bismuth 2-ethylhexanoate is used as an industrial lubricant and dessicant. It is also used for synthesizing of urethane foam, urethane coating and other urethane products.