Water, Reagent (Deionized water), ACS
Water, Reagent (Deionized water), ACS
Water, Reagent (Deionized water), ACS
Water, Reagent (Deionized water), ACS
Thermo Scientific Chemicals
Water, Reagent (Deionized water), ACS
CAS: 7732-18-5 | H2O | 18.015 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF036645.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialWater
CAS7732-18-5
ColorColorless
Melting Point0°C
Boiling Point100°C
View more
Used in conductometric titration, Synthesis of nanomaterials. Mostly it is used as solvent in industries and in all research fields.