Soda lime, ACS
Soda lime, ACS
Soda lime, ACS
Thermo Scientific Chemicals
Soda lime, ACS
CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol
Have Questions?
Catalog NumberQuantity
ALF036596.36500 g
Catalog number ALF036596.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSoda Lime
Physical Form4-8 Mesh
CAS8006-28-8
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
View more
Soda lime's ability to absorb carbon dioxide makes it valuable in medical and surgical professions. It is also used as powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil CO2 efflux upon absorption. It is also useful for direct measurement of dissolved CO2.