Azelaic acid, 98%
Azelaic acid, 98%
Azelaic acid, 98%
Thermo Scientific Chemicals
Azelaic acid, 98%
CAS: 123-99-9 | C9H16O4 | 188.223 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF036308.22
Price (MYR)
592.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
592.00
EA
Add to cart
Specifications
Chemical Name or MaterialAzelaic acid
CAS123-99-9
Melting Point∼102°C
Boiling Point286°C (100mmHg)
Recommended StorageAmbient temperatures
View more
Useful in lubrication and plasticizers. Azelaic acid is used to treat acne.