Samarium(III) oxide, 99.9% (metals basis)
Samarium(III) oxide, 99.9% (metals basis)
Samarium(III) oxide, 99.9% (metals basis)
Samarium(III) oxide, 99.9% (metals basis)
Thermo Scientific Chemicals
Samarium(III) oxide, 99.9% (metals basis)
CAS: 12060-58-1 | Sm2O3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF036278.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSamarium(III) oxide
Melting Point2300°C
CAS12060-58-1
ColorWhite
Recommended StorageAmbient temperatures
View more
The biocompatibility of natural samarium (III) oxide, which has previously been used for treatment in bone-related diseases was determined as a first step in its evaluation as a bone implant material. The rates and products of the purely heterogeneous oxidations of C2H6 (g) and C2H4 (g) on Sm2O3 in the presence of O2 (g) were investigated in a very low-pressure flow reactor by on-line molecular beam mass spectrometry, about 1000±100 K. Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. Also, it is used as a neutron absorber in control rods for nuclear power reactors. The oxide catalyzes dehydration of acyclic primary alcohols to aldehydes and ketones. Another use involves preparation of other samarium salts.