Ytterbium(III) oxide, 99.9% (REO)
Ytterbium(III) oxide, 99.9% (REO)
Ytterbium(III) oxide, 99.9% (REO)
Ytterbium(III) oxide, 99.9% (REO)
Thermo Scientific Chemicals
Ytterbium(III) oxide, 99.9% (REO)
CAS: 1314-37-0 | O3Yb2 | 394.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF036252.1425 g
Catalog number ALF036252.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialYtterbium oxide
Melting Point2227°C
CAS1314-37-0
Recommended StorageAmbient temperatures
Physical FormSintered Lump
View more
Ytterbium(III) oxide is being applied to numerous fiber amplifier and fiber optic technologies. High purity Ytterbium Oxide are widely applied as a doping agent for garnet crystals in lasers a important colorant in glasses and porcelain enamel glazes.. As Ytterbium Oxide has a significantly higher emissivity in the infrared range a higher radiant intensity is obtained with Ytterbium-based payloads.