Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Selenium powder, -200 mesh, 99.999% (metals basis)
Thermo Scientific Chemicals
Selenium powder, -200 mesh, 99.999% (metals basis)
CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF036208.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSelenium powder
CAS7782-49-2
Health Hazard 1H301+H331-H373
Health Hazard 2GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c