Calcium fluoride, ultra dry, 99.99% (metals basis)
Calcium fluoride, ultra dry, 99.99% (metals basis)
Calcium fluoride, ultra dry, 99.99% (metals basis)
Calcium fluoride, ultra dry, 99.99% (metals basis)
Thermo Scientific Chemicals
Calcium fluoride, ultra dry, 99.99% (metals basis)
CAS: 7789-75-5 | CaF2 | 78.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF035742.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Boiling Pointca 2500°C
Chemical Name or MaterialCalcium fluoride
Melting Point1403°C
CAS7789-75-5
ColorWhite
View more
Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.