Erbium(III) chloride, ultra dry, 99.9% (REO)
Erbium(III) chloride, ultra dry, 99.9% (REO)
Erbium(III) chloride, ultra dry, 99.9% (REO)
Erbium(III) chloride, ultra dry, 99.9% (REO)
Thermo Scientific Chemicals
Erbium(III) chloride, ultra dry, 99.9% (REO)
CAS: 10138-41-7 | Cl3Er | 273.609 g/mol
Have Questions?
Catalog NumberQuantity
ALF035739.065 g
Catalog number ALF035739.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Boiling Point1500°C
Chemical Name or MaterialErbium(III) chloride
Melting Point774°C
Name NoteUltra dry
CAS10138-41-7
View more
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ampouled under argon
RUO – Research Use Only