Hydrochloric acid, 0.01N Standardized Solution
Hydrochloric acid, 0.01N Standardized Solution
Hydrochloric acid, 0.01N Standardized Solution
Hydrochloric acid, 0.01N Standardized Solution
Thermo Scientific Chemicals
Hydrochloric acid, 0.01N Standardized Solution
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035648.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric acid
Name Note0.01N
CAS7647-01-0
CAS7732-18-5
Recommended StorageAmbient temperatures
View more
Hydrochloric acid, 0.01N standardized solution is used for volumetric analysis in laboratories.