Sodium thiosulfate, 0.01N Standardized Solution
Sodium thiosulfate, 0.01N Standardized Solution
Sodium thiosulfate, 0.01N Standardized Solution
Sodium thiosulfate, 0.01N Standardized Solution
Thermo Scientific Chemicals
Sodium thiosulfate, 0.01N Standardized Solution
CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
Have Questions?
Catalog NumberQuantity
ALF035643.K21 L
Catalog number ALF035643.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium thiosulfate
Name NoteStandardized Solution
CAS7772-98-7
Recommended StorageAmbient temperatures
EINECS Number231-867-5
View more
Sodium thiosulfate, 0.01N standardized solution is used in volumetric analysis to estimate the concentration of certain compounds in solution (hydrogen peroxide) as well as chlorine content in bleaching powder and water. It is used as a titrant in iodometry.