Hydrochloric acid, 1.0N Standardized Solution
Hydrochloric acid, 1.0N Standardized Solution
Hydrochloric acid, 1.0N Standardized Solution
Hydrochloric acid, 1.0N Standardized Solution
Thermo Scientific Chemicals
Hydrochloric acid, 1.0N Standardized Solution
CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035640.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialHydrochloric acid
CAS7647-01-0
Health Hazard 1H290-H315-H318
CAS7732-18-5
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P390
View more
Hydrochloric acid, 1.0N standardized solution is used for volumetric analysis in laboratories.