Sodium hydroxide, 20% w/v aq. soln.
Sodium hydroxide, 20% w/v aq. soln.
Sodium hydroxide, 20% w/v aq. soln.
Sodium hydroxide, 20% w/v aq. soln.
Thermo Scientific Chemicals
Sodium hydroxide, 20% w/v aq. soln.
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
Catalog number ALF035637.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide
CAS1310-73-2
Health Hazard 1H314-H335
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c