Sodium hydroxide, 2.0N Standardized Solution
Sodium hydroxide, 2.0N Standardized Solution
Sodium hydroxide, 2.0N Standardized Solution
Sodium hydroxide, 2.0N Standardized Solution
Thermo Scientific Chemicals
Sodium hydroxide, 2.0N Standardized Solution
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number ALF035631.K7
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
4 L
Request bulk or custom format
Specifications
Chemical Name or MaterialSodium hydroxide
CAS1310-73-2
Health Hazard 1H290-H314-H335
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c