Sodium chloride, 0.1N Standardized Solution
Sodium chloride, 0.1N Standardized Solution
Sodium chloride, 0.1N Standardized Solution
Sodium chloride, 0.1N Standardized Solution
Thermo Scientific Chemicals
Sodium chloride, 0.1N Standardized Solution
CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF035616.K2
Price (MYR)
438.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
438.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium chloride
CAS7647-14-5
Recommended StorageAmbient temperatures
EINECS Number231-598-3
TSCAYes
View more
Sodium chloride, 0.1N standardized solution is used for volumetric analysis in laboratories.