Specifications
Chemical Name or MaterialSulfuric acid
CAS7664-93-9
Health Hazard 1H290-H314-H335
Health Hazard 3P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c