Potassium hydrogen phthalate, 0.05N Standardized Solution
Potassium hydrogen phthalate, 0.05N Standardized Solution
Potassium hydrogen phthalate, 0.05N Standardized Solution
Potassium hydrogen phthalate, 0.05N Standardized Solution
Thermo Scientific Chemicals
Potassium hydrogen phthalate, 0.05N Standardized Solution
CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF035603.K2
Price (MYR)
423.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
423.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydrogen phthalate
Name NoteStandardized Solution
CAS877-24-7
Recommended StorageAmbient temperatures
EINECS Number212-889-4