Potassium dichromate, 0.25N Standardized Solution
Potassium dichromate, 0.25N Standardized Solution
Potassium dichromate, 0.25N Standardized Solution
Potassium dichromate, 0.25N Standardized Solution
Thermo Scientific Chemicals
Potassium dichromate, 0.25N Standardized Solution
CAS: 7778-50-9 | Cr2K2O7 | 294.182 g/mol
Have Questions?
Catalog NumberQuantity
ALF035587.K21 L
Catalog number ALF035587.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium dichromate
ColorOrange
Name NoteStandardized Solution
CAS7778-50-9
Health Hazard 1H315-H317-H319-H332-H334-H340-H350-H360FD-H373