Potassium iodate, 0.1N Standardized Solution
Potassium iodate, 0.1N Standardized Solution
Potassium iodate, 0.1N Standardized Solution
Potassium iodate, 0.1N Standardized Solution
Thermo Scientific Chemicals
Potassium iodate, 0.1N Standardized Solution
CAS: 7758-05-6 | IKO3 | 214.00 g/mol
Have Questions?
Catalog NumberQuantity
ALF035586.K21 L
Catalog number ALF035586.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium iodate
ColorColorless
Name NoteStandardized Solution
CAS7758-05-6
CAS7732-18-5
View more
Potassium iodate 0.1N solution is used as a standard solution in volumetric titrations. It is extensively used in analytical chemistry.