Potassium hexacyanoferrate(II), 0.1N Standardized Solution
Potassium hexacyanoferrate(II), 0.1N Standardized Solution
Potassium hexacyanoferrate(II), 0.1N Standardized Solution
Potassium hexacyanoferrate(II), 0.1N Standardized Solution
Thermo Scientific Chemicals
Potassium hexacyanoferrate(II), 0.1N Standardized Solution
CAS: 13943-58-3
Have Questions?
Catalog NumberQuantity
ALF035585.K21 L
Catalog number ALF035585.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium hexacyanoferrate(II)
ColorYellow
Name NoteStandardized Solution
CAS13943-58-3