Barium chloride, 10% w/v aq. soln.
Barium chloride, 10% w/v aq. soln.
Barium chloride, 10% w/v aq. soln.
Barium chloride, 10% w/v aq. soln.
Thermo Scientific Chemicals
Barium chloride, 10% w/v aq. soln.
CAS: 10361-37-2 | BaCl2 | 208.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF035584.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialBarium chloride
ColorColorless
CAS10361-37-2
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302