Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL
Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL
Thermo Scientific Chemicals
Nitrate, Ion chromatography standard solution, Specpure™ NO{3}∣- 1000μg/mL
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF035549.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialNitrate Ion chromatography standard solution
Name NoteNO3- 1000μg/mL
Recommended StorageAmbient temperatures
TSCAYes
Physical FormLiquid
View more
Nitrate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.