Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
Thermo Scientific Chemicals
Phosphate, Ion chromatography standard solution, Specpure™ PO4∣-^3 1000μg/mL
CAS: 7558-80-7 | H2NaO4P | 119.98 g/mol
Have Questions?
Catalog NumberQuantity
ALF035548.AE100 mL
Catalog number ALF035548.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialPhosphate
CAS7558-80-7
Recommended StorageAmbient temperatures
TSCAYes
Physical FormLiquid
View more
Phosphate, Ion chromatography standard solution is used as a standard solution in analytical chemistry and in ion chromatography. It is used for calibration of ion chromatography and other analytical techniques.