Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL
Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL
Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL
Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL
Thermo Scientific Chemicals
Aluminum, AAS standard solution, Specpure™ Al 1000μg/mL
CAS: 11145-27-0 | AlSi | 55.067 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF033557.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum
CAS11145-27-0
Health Hazard 1H290
Health Hazard 3P234-P390