Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Sulfur powder, sublimed, -100 mesh, 99.5%
Thermo Scientific Chemicals
Sulfur powder, sublimed, -100 mesh, 99.5%
CAS: 7704-34-9 | S | 32.06 g/mol
Have Questions?
Catalog NumberQuantity
ALF033394.36500 g
Catalog number ALF033394.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialSulphur
CAS7704-34-9
ColorYellow
Health Hazard 1H315-H319-H500
Health Hazard 3P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362