Carbon powder, activated
Carbon powder, activated
Carbon powder, activated
Carbon powder, activated
Thermo Scientific Chemicals
Carbon powder, activated
CAS: 7440-44-0 | C | 12.011 g/mol
Have Questions?
Catalog NumberQuantity
ALF033302.22100 g
Catalog number ALF033302.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCarbon powder
CAS7440-44-0
Health Hazard 1H252
Health Hazard 3P235+P410-P280
ColorBlack