Calcium carbonate, ACS, 99.0% min
Calcium carbonate, ACS, 99.0% min
Calcium carbonate, ACS, 99.0% min
Calcium carbonate, ACS, 99.0% min
Thermo Scientific Chemicals
Calcium carbonate, ACS, 99.0% min
CAS: 471-34-1 | CCaO3 | 100.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF033295.36500 g
Catalog number ALF033295.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCalcium carbonate
Melting Point800°C (decomposition)
Name NoteFor ACS analysis
CAS471-34-1
ColorWhite
View more
Calcium carbonate is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles show a useful application as an anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.