Acetic acid, glacial, 99+%
Acetic acid, glacial, 99+%
Acetic acid, glacial, 99+%
Acetic acid, glacial, 99+%
Thermo Scientific Chemicals
Acetic acid, glacial, 99+%
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
Catalog number ALF033252.K2
Price (MYR)
476.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
476.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcetic acid
Name Noteglacial
CAS64-19-7
Health Hazard 1H226-H314
Health Hazard 3P210-P233-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
View more
Good polar protic solvent, in its ester form it acts as a solvent for inks paints and coatings. Used in laboratory as a solvent for recrystallization, manufacture of camphor, cellulose acetate, polyvinyl acetate. It involves in Wagner-Meerwein rearrangement. Used for preparation of mono-lithium salt of 5-sulfoisophthalic acid.