Toluene, ACS, 99.5% min
Toluene, ACS, 99.5% min
Toluene, ACS, 99.5% min
Toluene, ACS, 99.5% min
Thermo Scientific Chemicals
Toluene, ACS, 99.5% min
CAS: 108-88-3 | C7H8 | 92.14 g/mol
Have Questions?
Catalog NumberQuantity
ALF031755.AP500 mL
Catalog number ALF031755.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialToluene
Melting Point°C to 93°C
CAS108-88-3
ColorColorless
Health Hazard 1H225-H304-H315-H319-H336-H361d-H373
View more
Toluene is used as solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.