Ethyl acetate, ACS, 99.5+%
Ethyl acetate, ACS, 99.5+%
Ethyl acetate, ACS, 99.5+%
Ethyl acetate, ACS, 99.5+%
Thermo Scientific Chemicals
Ethyl acetate, ACS, 99.5+%
CAS: 141-78-6 | C4H8O2 | 88.106 g/mol
Have Questions?
Catalog NumberQuantity
ALF031344.K21 L
Catalog number ALF031344.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialEthyl acetate
Melting Point-84°C
CAS141-78-6
ColorColorless
Health Hazard 1H225-H319-H336
View more
Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.