Tetramethylammonium perchlorate
Tetramethylammonium perchlorate
Tetramethylammonium perchlorate
Tetramethylammonium perchlorate
Thermo Scientific Chemicals
Tetramethylammonium perchlorate
CAS: 2537-36-2 | C4H12ClNO4 | 173.59 g/mol
Have Questions?
Catalog NumberQuantity
ALF030834.1425 g
Catalog number ALF030834.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTetramethylammonium perchlorate
CAS2537-36-2
Health Hazard 1H272-H315-H319-H335
Health Hazard 2GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
View more
Tetramethylammonium perchlorate acts as an intermediate in organic and chemical synthesis. It is used as a mobile phase in high performance liquid chromatographic analysis and in the quantitation of olanzapine clozapine and N-desmethylclozapine. It acts as a supporting electrolyte in the electrochemical reduction of electrochemical reduction of 1-bromodecanr, 1-iododecane,2-iodooctane and 2-bromooctane at mercury cathodes.