Ammonium benzoate, 99%
Ammonium benzoate, 99%
Ammonium benzoate, 99%
Ammonium benzoate, 99%
Thermo Scientific Chemicals
Ammonium benzoate, 99%
CAS: 1863-63-4 | C7H9NO2 | 139.154 g/mol
Have Questions?
Catalog NumberQuantity
ALF030753.22100 g
Catalog number ALF030753.22
Price (MYR)
215.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
215.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium benzoate
Melting Point192°C to 198°C (decomposition)
CAS1863-63-4
ColorWhite
Health Hazard 1H302-H315-H319-H335
View more
Ammonium benzoate is used predominantly in industries which acts as a corrosion inhibitors and anti-scaling agents . It is also used as paint additives and coating additives. Other consumer uses include cleaning and furnishing Care products, paints and coatings, personal care products, water treatment products. Ammonium benzoate is also used as chemical analysis reagents, in electronics industry, used for the production of electrolytic capacitors and also used in pharmaceutical industries. Used as adhesives, metal rust treatment and rubber additives.