Cerium, AAS standard solution, Specpure™ Ce 1000μg/mL
Cerium, AAS standard solution, Specpure™ Ce 1000μg/mL
Thermo Scientific Chemicals
Cerium, AAS standard solution, Specpure™ Ce 1000μg/mL
Have Questions?
Catalog NumberQuantity
ALF030520.AE100 mL
Catalog number ALF030520.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium AAS standard solution
Name NoteCe 1000μg/mL
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390