View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
FormatLiquid
For Use With (Application)Reagents and strips to measure 10-400ppm NH4+1 in aqueous solution
Storage RequirementsAmbient
TypeFully Miscible